Assay Details
Gene Symbol: IL17B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744355.3 | XP_002744401.1 | 2 - 3 | 390 | 74 | |
XM_017968827.1 | XP_017824316.1 | 3 - 4 | 470 | 74 | ||
XM_017968828.1 | XP_017824317.1 | 2 - 3 | 414 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
