Assay Details
| Gene Symbol: CD209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761676.3 | XP_002761722.3 | 3 - 4 | 254 | 64 | |
| XM_017968179.1 | XP_017823668.1 | 3 - 4 | 245 | 64 | ||
| GenBank mRNA | EU041929.1 | - | 3 - 4 | 186 | 64 | |
More Information
Set Membership: |
|

CD209
Genomic Map