Assay Details
Gene Symbol: ARL13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002761298.3 | XP_002761344.1 | 9 - 10 | 1462 | 68 | |
XM_008986440.2 | XP_008984688.1 | 9 - 10 | 1177 | 68 | ||
XM_017967201.1 | XP_017822690.1 | 8 - 9 | 1555 | 68 | ||
XM_017967203.1 | XP_017822692.1 | 8 - 9 | 1163 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
