Assay Details
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008980989.1 | XP_008979237.1 | 12 - 13 | 1594 | 82 | |
XM_008980990.2 | XP_008979238.1 | 11 - 12 | 1510 | 82 | ||
XM_017964462.1 | XP_017819951.1 | 11 - 12 | 1521 | 82 | ||
XM_017964463.1 | XP_017819952.1 | 12 - 13 | 1583 | 82 | ||
XM_017964464.1 | XP_017819953.1 | 12 - 13 | 1508 | 82 | ||
XM_017964465.1 | XP_017819954.1 | 11 - 12 | 1506 | 82 | ||
XM_017964466.1 | XP_017819955.1 | 12 - 13 | 1594 | 82 | ||
XM_017964467.1 | XP_017819956.1 | 12 - 13 | 1594 | 82 | ||
XM_017964468.1 | XP_017819957.1 | 12 - 13 | 1594 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
