Assay Details
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002759066.4 | XP_002759112.1 | 4 - 5 | 466 | 78 | |
XM_008982884.2 | XP_008981132.1 | 4 - 5 | 463 | 78 | ||
XM_008982886.2 | XP_008981134.1 | 3 - 4 | 307 | 78 | ||
XM_008982887.2 | XP_008981135.1 | 3 - 4 | 304 | 78 | ||
XM_017965450.1 | XP_017820939.1 | 4 - 5 | 666 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
