Assay Details
Gene Symbol: GUCY1A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745302.3 | XP_002745348.1 | 8 - 9 | 2104 | 84 | |
XM_002745303.4 | XP_002745349.1 | 8 - 9 | 2165 | 84 | ||
XM_008992555.2 | XP_008990803.1 | 8 - 9 | 2154 | 84 | ||
XM_008992556.2 | XP_008990804.1 | 8 - 9 | 2226 | 84 | ||
XM_008992557.2 | XP_008990805.1 | 7 - 8 | 2098 | 84 | ||
XM_008992559.2 | XP_008990807.1 | 9 - 10 | 2141 | 84 | ||
XM_017970181.1 | XP_017825670.1 | 7 - 8 | 3141 | 84 | ||
XM_017970182.1 | XP_017825671.1 | 8 - 9 | 2165 | 84 | ||
XM_017970183.1 | XP_017825672.1 | 9 - 10 | 2191 | 84 | ||
XM_017970184.1 | XP_017825673.1 | 8 - 9 | 2013 | 84 | ||
XM_017970185.1 | XP_017825674.1 | 8 - 9 | 2132 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
