Assay Details
Gene Symbol: CCDC82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754686.3 | XP_002754732.1 | 2 - 3 | 839 | 63 | |
XM_009007202.2 | XP_009005450.1 | 2 - 3 | 839 | 63 | ||
XM_017977676.1 | XP_017833165.1 | 2 - 3 | 1083 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
