Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755792.3 | XP_002755838.1 | 3 - 4 | 258 | 73 | |
XM_009008931.2 | XP_009007179.1 | 3 - 4 | 258 | 73 | ||
XM_017978625.1 | XP_017834114.1 | 3 - 4 | 258 | 73 | ||
XM_017978626.1 | XP_017834115.1 | 3 - 4 | 258 | 73 | ||
XM_017978627.1 | XP_017834116.1 | 3 - 4 | 258 | 73 | ||
XM_017978628.1 | XP_017834117.1 | 3 - 4 | 258 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
