Assay Details
| Gene Symbol: CX3CL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985928.2 | XP_008984176.1 | 2 - 3 | 296 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CX3CL1| Gene Symbol: CX3CL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985928.2 | XP_008984176.1 | 2 - 3 | 296 | 68 | |
Set Membership: |
|