Assay Details
Gene Symbol: BNIP3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756821.3 | XP_002756867.1 | 5 - 6 | 1237 | 73 | |
XM_008979087.2 | XP_008977335.1 | 5 - 6 | 970 | 73 | ||
XM_008979090.1 | XP_008977338.1 | 6 - 7 | 645 | 73 | ||
XM_017963678.1 | XP_017819167.1 | 6 - 7 | 732 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
