Assay Details
Gene Symbol: TMEM79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760098.3 | XP_002760144.1 | 3 - 4 | 1828 | 62 | |
XM_017966353.1 | XP_017821842.1 | 2 - 3 | 1203 | 62 | ||
XM_017966354.1 | XP_017821843.1 | 3 - 4 | 1977 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
