Assay Details
Gene Symbol: LOC100894590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008998060.2 | XP_008996308.1 | 2 - 3 | 158 | 118 | |
XM_008998061.2 | XP_008996309.1 | 2 - 3 | 573 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
