Assay Details
| Gene Symbol: ZFP91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755308.4 | XP_002755354.2 | 8 - 9 | 1207 | 92 | |
| XM_009008164.2 | XP_009006412.1 | 8 - 9 | 1210 | 92 | ||
| XM_009008165.2 | XP_009006413.1 | 7 - 8 | 1105 | 92 | ||
More Information
Set Membership: |
|

ZFP91
Genomic Map