Assay Details
Gene Symbol: RAB40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008997979.2 | XP_008996227.2 | 2 - 3 | 371 | 65 | |
XM_017972967.1 | XP_017828456.1 | 2 - 3 | 371 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
