Assay Details
Gene Symbol: DRAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009001866.2 | XP_009000114.1 | 5 - 6 | 603 | 72 | |
XM_009001867.2 | XP_009000115.1 | 4 - 5 | 458 | 72 | ||
XM_017975016.1 | XP_017830505.1 | 4 - 5 | 368 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
