Assay Details
Gene Symbol: INO80D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749678.4 | XP_002749724.2 | 11 | 7942 | 109 | |
XM_008999282.2 | XP_008997530.1 | 11 | 5876 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
