Assay Details
Gene Symbol: BCL7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753110.3 | XP_002753156.1 | 3 - 4 | 1716 | 56 | |
XM_002753111.2 | XP_002753157.1 | 3 - 4 | 476 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
