Assay Details
| Gene Symbol: LATS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748886.3 | XP_002748932.1 | 4 - 5 | 2370 | 59 | |
| XM_008998005.2 | XP_008996253.1 | 4 - 5 | 2370 | 59 | ||
| XM_017972994.1 | XP_017828483.1 | 2 - 3 | 1494 | 59 | ||
More Information
Set Membership: |
|

LATS2
Genomic Map