Assay Details
| Gene Symbol: NPR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745077.4 | XP_002745123.2 | 9 | 6783 | 70 | |
| XM_008992209.2 | XP_008990457.1 | 9 | 6786 | 70 | ||
More Information
Set Membership: |
|

NPR3
Genomic Map