Assay Details
Gene Symbol: SH3RF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008992468.2 | XP_008990716.1 | 4 - 5 | 935 | 87 | |
XM_017970151.1 | XP_017825640.1 | 4 - 5 | 998 | 87 | ||
XM_017970152.1 | XP_017825641.1 | 3 - 4 | 858 | 87 | ||
XM_017970153.1 | XP_017825642.1 | 3 - 4 | 861 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
