Assay Details
| Gene Symbol: LOC100414591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965542.1 | XP_017821031.1 | 2 - 3 | 273 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100414591| Gene Symbol: LOC100414591 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017965542.1 | XP_017821031.1 | 2 - 3 | 273 | 81 | |
Set Membership: |
|