Assay Details
Gene Symbol: RIPK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002746265.3 | XP_002746311.1 | 3 - 4 | 1025 | 76 | |
XM_008994039.2 | XP_008992287.1 | 4 - 5 | 980 | 76 | ||
XM_008994040.2 | XP_008992288.1 | 3 - 4 | 831 | 76 | ||
XM_017970943.1 | XP_017826432.1 | 3 - 4 | 1026 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
