Assay Details
Gene Symbol: RAB11FIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002756952.3 | XP_002756998.1 | 2 - 3 | 1021 | 71 | |
XM_008979321.2 | XP_008977569.1 | 2 - 3 | 1021 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
