Assay Details
| Gene Symbol: ELMOD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745334.4 | XP_002745380.1 | 9 | 3479 | 111 | |
| XM_008992676.2 | XP_008990924.1 | 9 | 3435 | 111 | ||
| XM_008992677.2 | XP_008990925.1 | 9 | 3420 | 111 | ||
More Information
Set Membership: |
|

ELMOD2
Genomic Map