Assay Details
Gene Symbol: BAHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753546.4 | XP_002753592.2 | 3 - 4 | 1904 | 86 | |
XM_009005556.1 | XP_009003804.1 | 3 - 4 | 1904 | 86 | ||
XM_017976924.1 | XP_017832413.1 | 3 - 4 | 1901 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
