Assay Details
| Gene Symbol: DOHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761594.4 | XP_002761640.2 | 2 - 3 | 463 | 80 | |
| XM_008986980.2 | XP_008985228.1 | 2 - 3 | 1116 | 80 | ||
| XM_008986981.2 | XP_008985229.1 | 2 - 3 | 467 | 80 | ||
| XM_008986984.2 | XP_008985232.1 | 2 - 3 | 1102 | 80 | ||
| XM_017967746.1 | XP_017823235.1 | 2 - 3 | 1111 | 80 | ||
More Information
Set Membership: |
|

DOHH
Genomic Map