Assay Details
Gene Symbol: STYX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753911.3 | XP_002753957.1 | 1 - 2 | 365 | 63 | |
XM_017977117.1 | XP_017832606.1 | 1 - 2 | 369 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
