Assay Details
Gene Symbol: DUPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009009678.2 | XP_009007926.1 | 2 - 3 | 330 | 67 | |
XM_009009679.2 | XP_009007927.1 | 3 - 4 | 351 | 67 |

More Information
Set Membership: |
![]() ![]() |
