Assay Details
Gene Symbol: ARHGAP24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745629.4 | XP_002745675.1 | 6 - 7 | 1189 | 71 | |
XM_002745630.4 | XP_002745676.1 | 4 - 5 | 3777 | 71 | ||
XM_008993039.2 | XP_008991287.1 | 6 - 7 | 857 | 71 | ||
XM_008993040.2 | XP_008991288.1 | 4 - 5 | 2747 | 71 | ||
XM_008993043.2 | XP_008991291.1 | 4 - 5 | 1357 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
