Assay Details
| Gene Symbol: CHRNB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757015.3 | XP_002757061.1 | 7 - 8 | 4829 | 92 | |
| XM_017963810.1 | XP_017819299.1 | 6 - 7 | 5078 | 92 | ||
| XM_017963811.1 | XP_017819300.1 | 7 - 8 | 4785 | 92 | ||
| XM_017963812.1 | XP_017819301.1 | 3 - 4 | 4441 | 92 | ||
| XM_017963813.1 | XP_017819302.1 | 5 - 6 | 4630 | 92 | ||
More Information
Set Membership: |
|

CHRNB3
Genomic Map