Assay Details
Gene Symbol: DCX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002763166.3 | XP_002763212.1 | 3 - 4 | 815 | 97 | |
XM_008989781.1 | XP_008988029.1 | 3 - 4 | 815 | 97 | ||
XM_017968670.1 | XP_017824159.1 | 3 - 4 | 939 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
