Assay Details
| Gene Symbol: SYT10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752372.3 | XP_002752418.1 | 4 - 5 | 1320 | 72 | |
| XM_017975995.1 | XP_017831484.1 | 4 - 5 | 1238 | 72 | ||
More Information
Set Membership: |
|

SYT10
Genomic Map