Assay Details
Gene Symbol: FAM234A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755727.3 | XP_002755773.2 | 3 - 4 | 501 | 61 | |
XM_009008816.1 | XP_009007064.1 | 3 - 4 | 546 | 61 | ||
XM_009008817.2 | XP_009007065.1 | 3 - 4 | 538 | 61 | ||
XM_009008818.1 | XP_009007066.1 | 3 - 4 | 489 | 61 | ||
XM_009008819.2 | XP_009007067.1 | 3 - 4 | 1062 | 61 | ||
XM_009008821.2 | XP_009007069.1 | 3 - 4 | 501 | 61 | ||
XM_017978592.1 | XP_017834081.1 | 2 - 3 | 360 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
