Assay Details
Gene Symbol: LOC100409482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008989360.2 | XP_008987608.1 | 2225 | 84 | ||
XM_017968514.1 | XP_017824003.1 | 2240 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
