Assay Details
Gene Symbol: LOC100389979 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745559.3 | XP_002745605.1 | 5 - 6 | 837 | 87 | |
XM_008992929.2 | XP_008991177.1 | 5 - 6 | 837 | 87 | ||
XM_017970403.1 | XP_017825892.1 | 5 - 6 | 837 | 87 | ||
GenBank mRNA | KM972570.1 | - | 5 - 6 | 567 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
