Assay Details
| Gene Symbol: LOC100389979 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745559.3 | XP_002745605.1 | 6 - 7 | 1102 | 75 | |
| XM_008992929.2 | XP_008991177.1 | 6 - 7 | 1102 | 75 | ||
| XM_017970403.1 | XP_017825892.1 | 6 - 7 | 1102 | 75 | ||
| GenBank mRNA | KM972570.1 | - | 6 - 7 | 832 | 75 | |
More Information
Set Membership: |
|

LOC100389979
Genomic Map