Assay Details
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760075.4 | XP_002760121.1 | 5 - 6 | 716 | 66 | |
XM_008984524.2 | XP_008982772.1 | 5 - 6 | 692 | 66 | ||
XM_008984525.2 | XP_008982773.1 | 4 - 5 | 485 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
