Assay Details
Gene Symbol: PSMB10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002761089.4 | XP_002761135.1 | 4 - 5 | 1140 | 72 | |
XM_008986090.2 | XP_008984338.1 | 4 - 5 | 712 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
