Assay Details
Gene Symbol: PAK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009005570.2 | XP_009003818.1 | 10 - 11 | 4450 | 59 | |
XM_009005571.1 | XP_009003819.1 | 10 - 11 | 2376 | 59 | ||
XM_009005572.2 | XP_009003820.1 | 9 - 10 | 4367 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
