Assay Details
Gene Symbol: RAB30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754744.3 | XP_002754790.1 | 4 - 5 | 599 | 62 | |
XM_009007348.2 | XP_009005596.1 | 4 - 5 | 2460 | 62 | ||
XM_009007350.2 | XP_009005598.1 | 3 - 4 | 2381 | 62 | ||
XM_017977748.1 | XP_017833237.1 | 3 - 4 | 1436 | 62 | ||
XM_017977749.1 | XP_017833238.1 | 2 - 3 | 1360 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
