Assay Details
Gene Symbol: LOC100407742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743536.3 | XP_002743582.1 | 5 - 6 | 648 | 93 | |
XM_009006649.2 | XP_009004897.1 | 5 - 6 | 648 | 93 | ||
XM_009006658.2 | XP_009004906.1 | 4 - 5 | 397 | 93 | ||
XM_009006666.2 | XP_009004914.1 | 4 - 5 | 2678 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
