Assay Details
| Gene Symbol: MINDY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976614.1 | XP_017832103.1 | 1375 | 119 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MINDY2| Gene Symbol: MINDY2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976614.1 | XP_017832103.1 | 1375 | 119 | ||