Assay Details
| Gene Symbol: GTF2E1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758763.3 | XP_002758809.1 | 3 - 4 | 814 | 76 | |
| XM_017965203.1 | XP_017820692.1 | 3 - 4 | 768 | 76 | ||
More Information
Set Membership: |
|

GTF2E1
Genomic Map