Assay Details
Gene Symbol: TIRAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009007089.2 | XP_009005337.1 | 3 - 4 | 365 | 74 | |
XM_017977637.1 | XP_017833126.1 | 2 - 3 | 1211 | 74 | ||
XM_017977638.1 | XP_017833127.1 | 2 - 3 | 1211 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
