Assay Details
Gene Symbol: TMEM155 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002806649.3 | XP_002806695.1 | 2 - 3 | 201 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002806649.3 transcripts.
Set Membership: |
![]() ![]() |
