Assay Details
| Gene Symbol: ATP5S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753882.4 | XP_002753928.1 | 1 - 2 | 145 | 103 | |
| XM_017977094.1 | XP_017832583.1 | 2 - 3 | 2249 | 103 | ||
| XM_017977095.1 | XP_017832584.1 | 1 - 2 | 147 | 103 | ||
More Information
Set Membership: |
|

ATP5S
Genomic Map