Assay Details
| Gene Symbol: IL2RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257218.1 | NP_001244147.1 | 1 - 2 | 90 | 60 | |
| GenBank mRNA | DQ520834.1 | - | 1 - 2 | 90 | 60 | |
More Information
Set Membership: |
|

IL2RA
Genomic Map