Assay Details
Gene Symbol: NUDT16L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755857.3 | XP_002755903.2 | 1 - 2 | 318 | 61 | |
XM_009009103.2 | XP_009007351.1 | 1 - 2 | 318 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
