Assay Details
| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746248.4 | XP_002746294.2 | 3 - 4 | 445 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LY86| Gene Symbol: LY86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746248.4 | XP_002746294.2 | 3 - 4 | 445 | 72 | |
Set Membership: |
|