Assay Details
| Gene Symbol: ZSCAN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970810.1 | XP_017826299.1 | 2 - 3 | 587 | 86 | |
| XM_017970811.1 | XP_017826300.1 | 2 - 3 | 587 | 86 | ||
| XM_017970812.1 | XP_017826301.1 | 2 - 3 | 587 | 86 | ||
| XM_017970813.1 | XP_017826302.1 | 2 - 3 | 587 | 86 | ||
More Information
Set Membership: |
|

ZSCAN12
Genomic Map